Ethyl-6-(4-cyanophenyl)-6-oxohexanoate ≥97%

Supplier: Matrix Scientific

087067-1G 087067-2G 087067-5G
102852-298EA 592.31 USD
102852-298 102852-300 102852-302
Ethyl-6-(4-cyanophenyl)-6-oxohexanoate ≥97%
Ethyl-6-(4-cyanophenyl)-6-oxohexanoate

Hazard: Irritant

Formula: C₁₅H₁₇NO₃
MDL Number: MFCD01320303
CAS Number: 951885-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR