4-[2-(Methylthio)phenyl]-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087049-1G 087049-2G 087049-5G
102852-190EA 490.77 USD
102852-190 102852-192 102852-194
4-[2-(Methylthio)phenyl]-4-oxobutyric acid ≥97%
4-[2-(Methylthio)phenyl]-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₁H₁₂O₃S
MDL Number: MFCD09801789
CAS Number: 951892-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR