8-(3,4,5-Trichlorophenyl)-8-oxooctanoic acid ≥97%
Supplier: Matrix Scientific
087048-1G
087048-2G
087048-5G
102852-184EA
476.92
USD
102852-184
102852-186
102852-188
8-(3,4,5-Trichlorophenyl)-8-oxooctanoic acid ≥97%
8-(3,4,5-Trichlorophenyl)-8-oxooctanoic acid
Hazard: Irritant
Formula:
C₁₄H₁₅Cl₃O₃ |
MDL Number:
MFCD09801776 CAS Number: 951891-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...