7-(3,4,5-Trichlorophenyl)-7-oxoheptanoic acid ≥97%

Supplier: Matrix Scientific

087047-1G 087047-2G 087047-5G
102852-178EA 476.92 USD
102852-178 102852-180 102852-182
7-(3,4,5-Trichlorophenyl)-7-oxoheptanoic acid ≥97%
7-(3,4,5-Trichlorophenyl)-7-oxoheptanoic acid

Hazard: Irritant

Formula: C₁₃H₁₃Cl₃O₃
MW: 323.61 g/mol
MDL Number: MFCD09801775
CAS Number: 951891-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR