4-(2-Chloro-6-fluorophenyl)-4-oxobutyric acid ≥97%
Supplier: Matrix Scientific
087045-1G
087045-2G
087045-5G
102852-166EA
490.77
USD
102852-166
102852-168
102852-170
4-(2-Chloro-6-fluorophenyl)-4-oxobutyric acid ≥97%
4-(2-Chloro-6-fluorophenyl)-4-oxobutyric acid
Hazard: Irritant
Formula:
C₁₀H₈ClFO₃ MW: 230.62 g/mol |
MDL Number:
MFCD09801762 CAS Number: 951891-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...