4'-Bromo-4-phenylbutyrophenone ≥97%

Supplier: Matrix Scientific

086864-1G 086864-2G 086864-5G
102851-086EA 672.31 USD
102851-086 102851-088 102851-090
4'-Bromo-4-phenylbutyrophenone ≥97%
4'-Bromo-4-phenylbutyrophenone

Hazard: Irritant

Formula: C₁₆H₁₅BrO
MDL Number: MFCD01851725
CAS Number: 111826-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR