Cyclohexyl-4-(2-methyl-1,3-dioxolan-2-yl)phenyl ketone ≥97%

Supplier: Matrix Scientific

086863-1G 086863-2G 086863-5G
102851-080EA 672.31 USD
102851-080 102851-082 102851-084
Cyclohexyl-4-(2-methyl-1,3-dioxolan-2-yl)phenyl ketone ≥97%
Cyclohexyl-4-(2-methyl-1,3-dioxolan-2-yl)phenyl ketone

Hazard: Irritant

Formula: C₁₇H₂₂O₃
MDL Number: MFCD19687271
CAS Number: 1346523-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR