2-Chloro-5-(4-cyanobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086664-1G 086664-2G 086664-5G
102849-886EA 795.38 USD
102849-886 102849-888 102849-890
2-Chloro-5-(4-cyanobenzoyl)pyridine ≥97%
2-Chloro-5-(4-cyanobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₃H₇ClN₂O
MDL Number: MFCD13153357
CAS Number: 1187171-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR