5-(3-Chlorobenzoyl)-2-chloropyridine ≥97%
Supplier: Matrix Scientific
086663-1G
086663-2G
086663-5G
102849-880EA
795.38
USD
102849-880
102849-882
102849-884
5-(3-Chlorobenzoyl)-2-chloropyridine ≥97%
5-(3-Chlorobenzoyl)-2-chloropyridine
Hazard: Irritant
Formula:
C₁₂H₇Cl₂NO |
MDL Number:
MFCD07775838 CAS Number: 1187168-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...