2-Chloro-5-(3-fluorobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086662-1G
086662-2G
086662-5G
102849-874EA
795.38
USD
102849-874
102849-876
102849-878
2-Chloro-5-(3-fluorobenzoyl)pyridine ≥97%
2-Chloro-5-(3-fluorobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₇ClFNO |
MDL Number:
MFCD07775852 CAS Number: 1187168-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...