2-Chloro-5-(4-methoxybenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086660-1G 086660-2G 086660-5G
102849-868EA 795.38 USD
102849-868 102849-870 102849-872
2-Chloro-5-(4-methoxybenzoyl)pyridine ≥97%
2-Chloro-5-(4-methoxybenzoyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂
MDL Number: MFCD07775854
CAS Number: 122601-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR