4-(3-Bromobenzoyl)-2-methylpyridine ≥97%

Supplier: Matrix Scientific

086658-1G 086658-2G 086658-5G
102849-856EA 795.38 USD
102849-856 102849-858 102849-860
4-(3-Bromobenzoyl)-2-methylpyridine ≥97%
4-(3-Bromobenzoyl)-2-methylpyridine

Hazard: Irritant

Formula: C₁₃H₁₀BrNO
MW: 276.14 g/mol
MDL Number: MFCD13153289
CAS Number: 1187169-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR