4-(2-Methoxybenzoyl)-2-methylpyridine ≥97%
Supplier: Matrix Scientific
086657-1G
086657-2G
086657-5G
102849-850EA
795.38
USD
102849-850
102849-852
102849-854
4-(2-Methoxybenzoyl)-2-methylpyridine ≥97%
4-(2-Methoxybenzoyl)-2-methylpyridine
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂ |
MDL Number:
MFCD13153273 CAS Number: 1187170-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...