5-(3-Methoxybenzoyl)-2-methylpyridine ≥97%

Supplier: Matrix Scientific

086654-1G 086654-2G 086654-5G
102849-832EA 795.38 USD
102849-832 102849-834 102849-836
5-(3-Methoxybenzoyl)-2-methylpyridine ≥97%
5-(3-Methoxybenzoyl)-2-methylpyridine

Hazard: Irritant

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD13153205
CAS Number: 1187163-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR