3-(4-Chlorobenzoyl)-4-methylpyridine ≥97%

Supplier: Matrix Scientific

086653-1G 086653-2G 086653-5G
102849-826EA 795.38 USD
102849-826 102849-828 102849-830
3-(4-Chlorobenzoyl)-4-methylpyridine ≥97%
3-(4-Chlorobenzoyl)-4-methylpyridine

Hazard: Irritant

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD13153150
CAS Number: 158978-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR