3-(2,4-Dimethoxybenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086651-1G 086651-2G 086651-5G
102849-814EA 795.38 USD
102849-814 102849-816 102849-818
3-(2,4-Dimethoxybenzoyl)pyridine ≥97%
3-(2,4-Dimethoxybenzoyl)pyridine

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD13152932
CAS Number: 83640-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR