3-(3-Methoxybenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086647-1G 086647-2G 086647-5G
102849-790EA 795.38 USD
102849-790 102849-792 102849-794
3-(3-Methoxybenzoyl)pyridine ≥97%
3-(3-Methoxybenzoyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD12153278
CAS Number: 260417-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR