2-(4-Chlorobenzoyl)-6-methoxypyridine ≥97%

Supplier: Matrix Scientific

086646-1G 086646-2G 086646-5G
102849-784EA 795.38 USD
102849-784 102849-786 102849-788
2-(4-Chlorobenzoyl)-6-methoxypyridine ≥97%
2-(4-Chlorobenzoyl)-6-methoxypyridine

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂
MDL Number: MFCD13152850
CAS Number: 1187171-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR