2-(2-Methoxybenzoyl)-3-methylpyridine ≥97%

Supplier: Matrix Scientific

086644-1G 086644-2G 086644-5G
102849-772EA 795.38 USD
102849-772 102849-774 102849-776
2-(2-Methoxybenzoyl)-3-methylpyridine ≥97%
2-(2-Methoxybenzoyl)-3-methylpyridine

Hazard: Irritant

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD13152768
CAS Number: 120280-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR