5-Methyl-2-(4-nitrobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086643-1G
086643-2G
086643-5G
102849-766EA
795.38
USD
102849-766
102849-768
102849-770
5-Methyl-2-(4-nitrobenzoyl)pyridine ≥97%
5-Methyl-2-(4-nitrobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₀N₂O₃ |
MDL Number:
MFCD13152661 CAS Number: 1187164-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...