Ethyl-5-(2-furanoyl)-2-thiophene carboxylate

Supplier: Matrix Scientific

086572-1G 086572-2G 086572-5G
102849-346EA 795.38 USD
102849-346 102849-348 102849-350
Ethyl-5-(2-furanoyl)-2-thiophene carboxylate
Ethyl-5-(2-furanoyl)-2-thiophene carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀O₄S
MDL Number: MFCD02260478
CAS Number: 890100-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR