3-(4-Methoxybenzoyl)-4-methylpyridine ≥97%

Supplier: Matrix Scientific

086566-1G 086566-2G 086566-5G
102849-310EA 795.38 USD
102849-310 102849-312 102849-314
3-(4-Methoxybenzoyl)-4-methylpyridine ≥97%
3-(4-Methoxybenzoyl)-4-methylpyridine

Hazard: Irritant

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD13153139
CAS Number: 1187167-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR