4-Chloro-2'-(thiomethyl)benzophenone ≥97%

Supplier: Matrix Scientific

086565-1G 086565-2G 086565-5G
102849-304EA 490.77 USD
102849-304 102849-306 102849-308
4-Chloro-2'-(thiomethyl)benzophenone ≥97%
4-Chloro-2'-(thiomethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₁₁ClOS
MDL Number: MFCD09801696
CAS Number: 951888-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR