4-Methoxy-4'-trifluoromethylbenzophenone ≥97%

Supplier: Matrix Scientific

086531-1G 086531-2G 086531-5G
102849-100EA 710.77 USD
102849-100 102849-102 102849-104
4-Methoxy-4'-trifluoromethylbenzophenone ≥97%
4-Methoxy-4'-trifluoromethylbenzophenone

Hazard: Irritant

Formula: C₁₅H₁₁F₃O₂
MDL Number: MFCD13152565
CAS Number: 6185-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR