trans-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086465-1G
086465-2G
086465-5G
102848-850EA
846.15
USD
102848-850
102848-852
102848-854
trans-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%
trans-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₅FO₃ Melting Pt: 173…176 °C |
MDL Number:
MFCD01319785 CAS Number: 736136-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...