trans-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086464-1G 086464-2G 086464-5G
102848-844EA 846.15 USD
102848-844 102848-846 102848-848
trans-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid ≥95%
trans-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₃
Melting Pt: 197…200 °C
MDL Number: MFCD01319775
CAS Number: 736136-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR