cis-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086463-1G 086463-2G 086463-5G
102848-838EA 998.46 USD
102848-838 102848-840 102848-842
cis-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%
cis-4-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₅FO₃
MDL Number: MFCD01311335
CAS Number: 735270-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR