cis-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086462-1G 086462-2G 086462-5G
102848-832EA 998.46 USD
102848-832 102848-834 102848-836
cis-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid ≥95%
cis-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₃
MDL Number: MFCD01311353
CAS Number: 735269-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR