trans-2-(2-Thiomethylbenzoyl)cyclohexane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086457-1G
086457-10G
102848-802EA
795.38
USD
102848-802
102848-806
trans-2-(2-Thiomethylbenzoyl)cyclohexane-1-carboxylic acid ≥95%
trans-2-(2-Thiomethylbenzoyl)cyclohexane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₈O₃S Melting Pt: 151…154 °C |
MDL Number:
MFCD07698700 CAS Number: 1134611-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...