cis/trans-3-Carbomethoxycyclohexane-1-carboxylic acid ≥97%
Supplier: Matrix Scientific
086450-1G
086450-10G
102848-760EA
490.77
USD
102848-760
102848-764
cis/trans-3-Carbomethoxycyclohexane-1-carboxylic acid ≥97%
cis/trans-3-Carbomethoxycyclohexane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₈H₂₈O₈ |
MDL Number:
MFCD07698696 CAS Number: 733742-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...