Ethyl-4-(2-tetrahydro-2H-pyranoxy)benzoylformate ≥97%
Supplier: Matrix Scientific
086414-1G
086414-5G
102848-544EA
530.77
USD
102848-544
102848-548
Ethyl-4-(2-tetrahydro-2H-pyranoxy)benzoylformate ≥97%
Ethyl-4-(2-tetrahydro-2H-pyranoxy)benzoylformate
Hazard: Irritant
Formula:
C₁₅H₁₈O₅ |
MDL Number:
MFCD09801439 CAS Number: 951888-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...