Ethyl-4-methoxy-2-methylbenzoylformate ≥97%

Supplier: Matrix Scientific

086406-1G 086406-5G
102848-496EA 530.77 USD
102848-496 102848-500
Ethyl-4-methoxy-2-methylbenzoylformate ≥97%
Ethyl-4-methoxy-2-methylbenzoylformate

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MDL Number: MFCD09801430
CAS Number: 951888-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR