Ethyl-2-isopropylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2-isopropylphenyl)-2-oxoacetate

086404-1G 086404-5G
102848-484EA 800 USD
102848-484 102848-488
Ethyl-2-isopropylbenzoylformate ≥97%
Ethyl-2-isopropylbenzoylformate

Hazard: Irritant

Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
MDL Number: MFCD09801428
CAS Number: 951888-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR