Ethyl-3-fluoro-4-phenylbenzoylformate ≥97%

Supplier: Matrix Scientific

086396-1G 086396-5G
102848-436EA 530.77 USD
102848-436 102848-440
Ethyl-3-fluoro-4-phenylbenzoylformate ≥97%
Ethyl-3-fluoro-4-phenylbenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₆H₁₃FO₃
MDL Number: MFCD09801421
CAS Number: 951888-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR