Ethyl-2-ethylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2-Ethylphenyl)-2-oxoacetate

086393-1G 086393-5G
102848-418EA 530.77 USD
102848-418 102848-422
Ethyl-2-ethylbenzoylformate ≥97%
Ethyl-2-ethylbenzoylformate

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD09801418
CAS Number: 951888-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR