Ethyl-3-(1,3-dioxan-2-yl)-4-methoxybenzoylformate ≥97%

Supplier: Matrix Scientific

086391-1G 086391-5G
102848-406EA 530.77 USD
102848-406 102848-410
Ethyl-3-(1,3-dioxan-2-yl)-4-methoxybenzoylformate ≥97%
Ethyl-3-(1,3-dioxan-2-yl)-4-methoxybenzoylformate

Hazard: Irritant

Formula: C₁₅H₁₈O₆
MDL Number: MFCD09801416
CAS Number: 951888-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR