Ethyl-3-(1,3-dioxan-2-yl)-4-fluorobenzoylformate ≥97%
Supplier: Matrix Scientific
086389-1G
086389-5G
102848-394EA
530.77
USD
102848-394
102848-398
Ethyl-3-(1,3-dioxan-2-yl)-4-fluorobenzoylformate ≥97%
Ethyl-3-(1,3-dioxan-2-yl)-4-fluorobenzoylformate
Hazard: Irritant
Formula:
C₁₄H₁₅FO₅ |
MDL Number:
MFCD09801414 CAS Number: 951888-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...