Ethyl-3-(N,N-dimethylamino)benzoylformate ≥97%

Supplier: Matrix Scientific

086385-1G 086385-5G
102848-370EA 530.77 USD
102848-370 102848-374
Ethyl-3-(N,N-dimethylamino)benzoylformate ≥97%
Ethyl-3-(N,N-dimethylamino)benzoylformate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD09801410
CAS Number: 951888-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR