Ethyl-2,3-dimethylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2,3-dimethylphenyl)-2-oxoacetate

086383-1G 086383-5G
102848-358EA 387.69 USD
102848-358 102848-362
Ethyl-2,3-dimethylbenzoylformate ≥97%
Ethyl-2,3-dimethylbenzoylformate

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD09801407
CAS Number: 951888-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR