Ethyl-4-cyano-3-fluorobenzoylformate ≥97%

Supplier: Matrix Scientific

086373-1G 086373-5G
102848-298EA 530.77 USD
102848-298 102848-302
Ethyl-4-cyano-3-fluorobenzoylformate ≥97%
Ethyl-4-cyano-3-fluorobenzoylformate

Hazard: Irritant

Formula: C₁₁H₈FNO₃
MDL Number: MFCD09801401
CAS Number: 951888-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR