Ethyl-2-cyano-3-fluorobenzoylformate ≥97%

Supplier: Matrix Scientific

086371-1G 086371-5G
102848-286EA 833.85 USD
102848-286 102848-290
Ethyl-2-cyano-3-fluorobenzoylformate ≥97%
Ethyl-2-cyano-3-fluorobenzoylformate

Hazard: Irritant

Formula: C₁₁H₈FNO₃
Melting Pt: 51…53 °C
MDL Number: MFCD09801399
CAS Number: 951888-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR