5'-Bromo-3-methyl[2,2']bipyridinyl ≥97%
Supplier: Matrix Scientific
086315-1G
086315-5G
102847-962EA
592.31
USD
102847-962
102847-966
5'-Bromo-3-methyl[2,2']bipyridinyl ≥97%
5'-Bromo-3-methyl[2,2']bipyridinyl
Hazard: Irritant
Formula:
C₁₁H₉BrN₂ MW: 249.12 g/mol |
MDL Number:
MFCD13152520 CAS Number: 1159281-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...