5-(2-Methylpyridin-4-yl)thiophene-2-carboxylic acid ethyl ester ≥97%

Supplier: Matrix Scientific

086313-1G 086313-5G
102847-950EA 592.31 USD
102847-950 102847-954
5-(2-Methylpyridin-4-yl)thiophene-2-carboxylic acid ethyl ester ≥97%
5-(2-Methylpyridin-4-yl)thiophene-2-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₃H₁₃NO₂S
MW: 247.32 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD13152511
CAS Number: 1187168-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR