Ethyl-5-(4-methyl-3-pyridyl)-2-furoate ≥97%
Supplier: Matrix Scientific
086311-1G
086311-5G
102847-938EA
592.31
USD
102847-938
102847-942
Ethyl-5-(4-methyl-3-pyridyl)-2-furoate ≥97%
Ethyl-5-(4-methyl-3-pyridyl)-2-furoate
Hazard: Irritant
Formula:
C₁₃H₁₃NO₃ MW: 231.25 g/mol Melting Pt: 37…40 °C |
MDL Number:
MFCD13152496 CAS Number: 1187163-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...