4-(2-Cyclohexenyl)isoquinoline ≥97%

Supplier: Matrix Scientific

086309-1G 086309-5G
102847-926EA 592.31 USD
102847-926 102847-930
4-(2-Cyclohexenyl)isoquinoline ≥97%
4-(2-Cyclohexenyl)isoquinoline

Hazard: Irritant

Formula: C₁₅H₁₅N
Boiling Pt: 165 °C (1 Torr)
MDL Number: MFCD13152476
CAS Number: 111678-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR