3-(2-Methoxy-5-methylphenyl)-2-methyl-1-propene ≥97%

Supplier: Matrix Scientific

086279-1G 086279-5G
102847-746EA 387.69 USD
102847-746 102847-750
3-(2-Methoxy-5-methylphenyl)-2-methyl-1-propene ≥97%
3-(2-Methoxy-5-methylphenyl)-2-methyl-1-propene

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD07775073
CAS Number: 951894-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR