3-(4-Bromo-2-fluorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-fluoro-1-(2-propen-1-yl)benzene
086277-1G
086277-5G
102847-734EA
387.69
USD
102847-734
102847-738
3-(4-Bromo-2-fluorophenyl)-1-propene ≥97%
3-(4-Bromo-2-fluorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₈BrF MW: 215.07 g/mol |
MDL Number:
MFCD09801298 CAS Number: 951894-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...