3-(2,4-Difluorophenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086270-1G 086270-5G
102847-692EA 387.69 USD
102847-692 102847-696
3-(2,4-Difluorophenyl)-1-propene ≥97%
3-(2,4-Difluorophenyl)-1-propene

Hazard: Irritant

Formula: C₉H₈F₂
MDL Number: MFCD09801255
CAS Number: 951893-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR