4-(2,5-Dimethoxyphenyl)-2-methyl-1-butene ≥97%

Supplier: Matrix Scientific

086256-1G 086256-5G
102847-608EA 592.31 USD
102847-608 102847-612
4-(2,5-Dimethoxyphenyl)-2-methyl-1-butene ≥97%
4-(2,5-Dimethoxyphenyl)-2-methyl-1-butene

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD09801186
CAS Number: 951891-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR