4-(2,3-Dimethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086249-1G 086249-5G
102847-572EA 592.31 USD
102847-572 102847-576
4-(2,3-Dimethoxyphenyl)-1-butene ≥97%
4-(2,3-Dimethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD09801168
CAS Number: 951890-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR